MilliporeSigma
  • A "jellyfish" shaped green emitting gallium(III)-containing metallomesogen.
A "jellyfish" shaped green emitting gallium(III)-containing metallomesogen.
Chemical communications (Cambridge, England) (2008-05-09)
Daniela Pucci, Iolinda Aiello, Anna Bellusci, Alessandra Crispini, Irene De Franco, Mauro Ghedini, Massimo La Deda
ABSTRACT
The synthesis of the first gallium(III)-based liquid crystal has been achieved grafting around the metal centre two chelating 2-methylquinolin-8-olate anions and one monodentate 3,4,5-tris(hexadecyloxy)benzoyloxy ligand, allowing the resulting complex to be a soft luminescent material with the typical high quantum yield of pentacordinated gallium species.
Sigma-Aldrich
Quinaldine, ≥95.0% (GC)
Supelco
Quinaldine, analytical standard, ≥90% (GC)