MilliporeSigma
  • Managing acute on chronic respiratory failure: a guide to non-invasive ventilation.
Managing acute on chronic respiratory failure: a guide to non-invasive ventilation.
British journal of hospital medicine (London, England : 2005) (2008-09-12)
J J P Goldring, J A Wedzicha
ABSTRACT
Acute-on-chronic respiratory failure is becoming an increasingly common medical emergency, mostly as a result of exacerbations of chronic obstructive pulmonary disease. It is associated with high mortality rates and so practising the best evidence-based management is vital.
Product Description
Doxapram hydrochloride, European Pharmacopoeia (EP) Reference Standard