MilliporeSigma
An oral treatment for lead toxicity.
Postgraduate medical journal (1991-01-01)
P S Thomas, C Ashton
ABSTRACT
Chronic lead poisoning has traditionally been treated by parenteral agents. We present a case where a comparison of ethylene diaminetetra-acetic acid was made with 2,3-dimethyl succinic acid (DMSA) which has the advantage of oral administration associated with little toxicity and appeared to be at least as efficacious.
Sigma-Aldrich
2,3-Dimethylsuccinic acid, ≥99%