MilliporeSigma
  • Differences in in vitro cerebellar neuronal responses to hypoxia in eider ducks, chicken and rats.
Differences in in vitro cerebellar neuronal responses to hypoxia in eider ducks, chicken and rats.
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology (2009-09-26)
Stian Ludvigsen, Lars P Folkow
ABSTRACT
Ducks are well-known to be more tolerant to asphyxia than non-diving birds, but it is not known if their defences include enhanced neuronal hypoxia tolerance. To test this, we compared extracellular recordings of spontaneous activity in the Purkinje cell layer of 400 mum thick isolated cerebellar slices from eider ducks, chickens and rats, before, during and after 60 min hypoxia (95%N(2)-5%CO(2)) or chemical anoxia (hypoxia + 2 mM NaCN). Most slices rapidly lost activity in hypoxia, with or without recovery after rinse and return to normoxia (95%O(2)-5%CO(2)), but some maintained spontaneous activity throughout the insult. Proportions of 'surviving' (i.e. recovering or active) duck slices were significantly higher than for chickens in anoxia, and relative activity levels were higher for ducks than for chickens during hypoxia, anoxia and recovery. Survival of rat slices was significantly poorer than for birds under all conditions. Results suggest that (1) duck cerebellar neurons are intrinsically more hypoxia-tolerant than chicken neurons; (2) avian neurons are more hypoxia-tolerant than rat neurons, and (3) the enhanced hypoxic tolerance of duck neurons largely depended on efficient anaerobiosis since it mainly manifested itself in chemical anoxia. Mechanisms underlying the observed differences in neuronal hypoxic responses remain to be elucidated.
Sigma-Aldrich
Sodium cyanate, 96%