MilliporeSigma
  • Structural requirements for repellency: norsesquiterpenes and sesquiterpenoid derivatives of nootkatone against the Formosan subterranean termite (Isoptera: Rhinotermitidae).
Structural requirements for repellency: norsesquiterpenes and sesquiterpenoid derivatives of nootkatone against the Formosan subterranean termite (Isoptera: Rhinotermitidae).
Pest management science (2010-07-06)
Betty C R Zhu, Gregg Henderson, Anne M Sauer, William Crowe, Roger A Laine
ABSTRACT
Research has shown that the family of grapefruit flavors called nootkatones have significant repellant and toxic effects to Formosan subterranean termites (Coptotermes formosanus Shiraki). Nineteen synthetic nootkatone derivatives, along with three commercially available nootkatone derivatives, were tested for repellent activity against C. formosanus by a choice assay in a petri dish with a two-step triage procedure. Based on the repellency threshold value, the relationships between structure and activity are discussed. Four derivatives of nootkatone have very high repellency and toxicity to C. formosanus, 9 times the potency of the primary compound nootkatone. Four other compounds have between 2 and 3 times the repellency of nootkatones, and three compounds are equal in their repellency to nootkatone.
Sigma-Aldrich
(+)-Nootkatone, ≥99.0% (GC)