MilliporeSigma
  • Comparison of cytochrome P450 3A enzymes in cynomolgus monkeys and humans.
Comparison of cytochrome P450 3A enzymes in cynomolgus monkeys and humans.
Drug metabolism and pharmacokinetics (2010-09-04)
Kazuhide Iwasaki, Norie Murayama, Ryo Koizumi, Yasuhiro Uno, Hiroshi Yamazaki
ABSTRACT
Drug metabolizing activities of cytochromes P450 (P450s, or CYPs) 3A4 and 3A5 in liver microsomes from the cynomolgus monkey [Macaca fascicularis (mf)] were investigated and compared with those of human P450 3A enzymes. Low activities for dealkylation of ethoxyresorufin and pentoxyresorufin were seen in recombinant monkey mfCYP3A4 and mfCYP3A5 and in recombinant human CYP3A4 and CYP3A5 expressed in bacterial membranes. Hydroxylation activities of mfCYP3A4 and mfCYP3A5 toward coumarin, paclitaxel, diclofenac, flurbiprofen, and S-mephenytoin were below detectable levels, as was also true for CYP3A4 and CYP3A5. Monkey mfCYP3A5 and mfCYP3A4 were highly active in bufuralol 1'-hydroxylation. mfCYP3A5 was efficient at dextromethorphan O-demethylation, although human CYP3A5 was unable to catalyze this reaction. Apparent bufuralol 1'-hydroxylation and dextromethorphan O-demethylation activities of monkey liver microsomes were higher than those of human liver microsomes, possibly because of contributions of mfCYP3A5 to these P450 2D-dependent drug oxidations. mfCYP3A5 and CYP3A5 catalyzed midazolam 1'-hydroxylation at a low substrate concentration more efficiently than the corresponding CYP3A4. mfCYP3A5 had higher testosterone 6beta-hydroxylase activity than mfCYP3A4, but the reverse relationship was observed in oxidation of nifedipine and hydroxylation of dexamethasone. These results demonstrate that monkey P450 3A enzymes have similar substrate selectivity to that of human P450 3A enzymes, but exhibit wider substrate selectivity toward P450 2D substrates.
Sigma-Aldrich
(±)-Bufuralol hydrochloride