MilliporeSigma
  • Exercise-induced stress inhibits both the induction and elicitation phases of in vivo T-cell-mediated immune responses in humans.
Exercise-induced stress inhibits both the induction and elicitation phases of in vivo T-cell-mediated immune responses in humans.
Brain, behavior, and immunity (2011-03-03)
Adam D Harper Smith, Sarah L Coakley, Mark D Ward, Andrew W Macfarlane, Peter S Friedmann, Neil P Walsh
ABSTRACT
Little is known about the influence of exercise on induction and elicitation phases of in vivo immunity in humans. We used experimental contact-hypersensitivity, a clinically relevant in vivo measure of T cell-mediated immunity, to investigate the effects of exercise on induction and elicitation phases of immune responses to a novel antigen. The effects of 2 h-moderate-intensity-exercise upon the induction (Study One) and elicitation of in vivo immune memory (Study Two) to diphenylcyclopropenone (DPCP) were examined. Study One: matched, healthy males were randomly-assigned to exercise (N=16) or control (N=16) and received a primary DPCP exposure (sensitization), 20 min after either 2 h running at 60% V O(2peak) (EX) or 2 h seated rest (CON). Four weeks later, participants received a low, dose-series DPCP challenge (elicitation) on their upper inner arm, which was read at 24 and 48 h as clinical score, oedema (skinfold thickness) and redness (erythema). Study Two: pilot; 13 healthy males were sensitized to DPCP. Elicitation challenges were repeated every 4 weeks until responses reached a reproducible plateau. Then, N=9 from the pilot study completed both EX and CON trials in a randomized order. Elicitation challenges were applied and evaluated as in Study One. Results demonstrate that exercise-induced stress significantly impairs both the induction (oedema -53% at 48 h; P<0.001) and elicitation (oedema -19% at 48 h; P<0.05) phases of the in vivo T-cell-mediated immune response. These findings demonstrate that prolonged moderate-intensity exercise impairs the induction and elicitation phases of in vivo T-cell-mediated immunity. Moreover, the induction component of new immune responses appears more sensitive to systemic-stress-induced modulation than the elicitation component.
Sigma-Aldrich
Diphenylcyclopropenone, 98%