MilliporeSigma
IR spectroscopy on jet-cooled isolated two-station rotaxanes.
The journal of physical chemistry. A (2011-04-29)
Anouk M Rijs, Euan R Kay, David A Leigh, Wybren Jan Buma
ABSTRACT
High-resolution IR spectroscopy has been employed to study isolated, switchable [2]rotaxanes. IR absorption spectra of two-station rotaxanes, their separate thread, and macrocycle components, as well as those of the individual stations incorporated into the thread, have been measured in the 1800-1000 cm(-1) region. These spectra have been fully analyzed, aided by quantum chemical predictions of the IR spectra. From these analyses, a comprehensive picture emerges of the conformational structure and binding interactions between the mechanically interlocked components of the rotaxane.
Sigma-Aldrich
Succinamide, 98%