MilliporeSigma
Two new phthalate derivatives from Nepeta clarkei (Labiatae).
Journal of Asian natural products research (2012-01-24)
Javid Hussain, Najeeb Ur Rehman, Hidayat Hussain, Liaqat Ali, Ahmed Al-Harrasi
ABSTRACT
Two new phthalates bis(2-ethylundecyl)phthalate and bis(2-ethyltridecyl)phthalate have been isolated from the chloroform-soluble portion of the whole plant of Nepeta clarkei along with one known compound β-amyrin. The structures of the two new compounds and β-amyrin were assigned on the basis of their ¹H and ¹³C NMR spectra including 2D NMR techniques such as COSY, HMQC, and HMBC experiments and compared with the literature data.
Product Description
Supelco
α-Amyrin, analytical standard
Supelco
β-Amyrin, analytical standard