MilliporeSigma
  • Allium ursinum L.: bioassay-guided isolation and identification of a galactolipid and a phytosterol exerting antiaggregatory effects.
Allium ursinum L.: bioassay-guided isolation and identification of a galactolipid and a phytosterol exerting antiaggregatory effects.
Pharmacology (2012-04-18)
Dina Sabha, Bahi Hiyasat, Kristina Grötzinger, Lothar Hennig, Franziska Schlegel, Friedrich-Wilhelm Mohr, Hans Wilhelm Rauwald, Stefan Dhein
ABSTRACT
We wanted to investigate the possible antithrom botic effects and elucidate the chemical identity of the active principles involved in inhibitory effects against adenosine diphosphate(ADP)-induced aggregation of human platelets by wild garlic, Allium ursinum L. For this purpose, a bioassay-guided isolation procedure was used followed by spectrometric identification of pure active compounds. For the bioassay, blood was taken from healthy human volunteers and platelet-rich plasma was prepared for turbidimetric platelet aggregation tests. Platelet-rich plasma, stimulated with 20 μ mol/l of ADP, was treated with extracts of different polarities, fractions and isolated single compounds from A. ursinum. The extracts were investigated by thin-layer chromatography(TLC), HPLC, mass spectroscopy, electrospray ionization mass spectrometry (ESI-MS) and 1/2-dimensional (1)H/(13) C-nuclear magnetic resonance (NMR) spectroscopic techniques. Fresh A. ursinum leaves were extracted with ethanol, which was the potent form that effectively inhibited ADP-induced aggregation of human platelets. Thisethanolic extract was subjected to liquid-liquid partition. Whilst the aqueous phase, containing the moiety of cysteine sulphoxide and thiosulphinate derivatives, showed only weak activity on platelet aggregation, the ethyl-acetate and particularly the chloroform partitions showed the high estaggregation-inhibiting potency. Thus, in our bioassay, the effects of alliins/allicins could be neglected. The chloroform phase, possessing the strongest activity, was separated into 28 fractions by gradient-elution open column chromatography on silica gel. The most active fractions 11–17 were separated again, yielding 10 subfractions. This afforded 1,2-di-O-α-linolenoyl-3-O-β-D-galactopyranosyl-sn-glycerol and β-sitosterol-3-O-β-D-glucopyranoside, the structures of which were determined by ESI-MS and 1/2-dimensional (1)H/(13) CNMR spectroscopic techniques. Furthermore, the minute amounts of volatile oil of A. ursinum leaves obtained by steam distillation according to Ph. Eur. could be evaluated asa third aggregation-inhibiting principle. In our study, for the first time, 2 active, non-sulphur-containing constituents of wild garlic, namely a galactolipid and a phytosterol,could be identified exhibiting inhibitory action on ADP-induced aggregation in human blood platelets. As a major constituent, the galactolipid, 1,2-di-O-α-linolenoyl-3-O-β-D-galactopyranosyl-sn-glycerol, not yet found in Allium sp., appears as a new, highly useful marker substance for A. ursinum drugs, or their pharmaceutical or food preparations,as shown by our orientating TLC analyses.
Product Description
Supelco
β-Sitosterol β-D-glucoside, analytical standard