MilliporeSigma
  • Evolution of microglial activation in ischaemic core and peri-infarct regions after stroke: a PET study with the TSPO molecular imaging biomarker [((11))C]vinpocetine.
Evolution of microglial activation in ischaemic core and peri-infarct regions after stroke: a PET study with the TSPO molecular imaging biomarker [((11))C]vinpocetine.
Journal of the neurological sciences (2012-07-31)
Balázs Gulyás, Miklós Tóth, Martin Schain, Anu Airaksinen, Adám Vas, Konstantinos Kostulas, Per Lindström, Jan Hillert, Christer Halldin
ABSTRACT
Although there is increasing evidence for microglial activation after an ischaemic stroke in the infarct core and the peri-infarct region, the "evolution" of the process in stroke patients is poorly known. Using PET and [((11))C]vinpocetine, we measured the regional changes of TSPO in the brain of nine ischaemic stroke patients up to 14weeks after the insult. Already a week after stroke there was an increased radioligand uptake, indicating the up-regulation of TSPO and the presence of activated microglia, in both the ischaemic core and the peri-infarct zone. This increased activation showed a steady decrease with post stroke time. The proportion between %SUV values in the peri-infarct zone and the ischaemic core increased with time. There were no time-dependent TSPO activity changes in other regions, not affected directly by the stroke. The present observations demonstrate that increased regional microglia activation, as a consequence of stroke, can be visualised with PET, using the TSPO molecular imaging biomarker [((11))C]vinpocetine. The evolution of this microglial activation shows a time dependent decrease the gradient of which is different between the peri-infarct zone and the ischaemic core. The findings indicate an increased microglial activation in the peri-stroke region for several weeks after the insult.
Sigma-Aldrich
Vinpocetine, ≥98%, solid