MilliporeSigma
  • Enantioselectivity in visible light-induced, singlet oxygen [2+4] cycloaddition reactions (type II photooxygenations) of 2-pyridones.
Enantioselectivity in visible light-induced, singlet oxygen [2+4] cycloaddition reactions (type II photooxygenations) of 2-pyridones.
Chemical communications (Cambridge, England) (2012-09-11)
Christian Wiegand, Eberhardt Herdtweck, Thorsten Bach
ABSTRACT
3-Substituted 2-pyridones were enantioselectively (68-90% ee) converted into the respective 3-hydroxypyridine-2,6-diones by a sequence consisting of a template-mediated type II photooxygenation and an acid-catalysed rearrangement.
Sigma-Aldrich
2-Hydroxypyridine, 97%
Levetiracetam impurity C, European Pharmacopoeia (EP) Reference Standard