MilliporeSigma
  • Identification and validation of a novel lead compound targeting 4-diphosphocytidyl-2-C-methylerythritol synthetase (IspD) of mycobacteria.
Identification and validation of a novel lead compound targeting 4-diphosphocytidyl-2-C-methylerythritol synthetase (IspD) of mycobacteria.
European journal of pharmacology (2012-09-15)
Peng Gao, Yanhui Yang, Chunling Xiao, Yishuang Liu, Maoluo Gan, Yan Guan, Xueqin Hao, Jianzhou Meng, Shuang Zhou, Xiaojuan Chen, Jiafei Cui
ABSTRACT
Tuberculosis is a serious threat to world-wide public health usually caused in humans by Mycobacterium tuberculosis (M. tuberculosis). It exclusively utilizes the methylerythritol phosphate (MEP) pathway for biosynthesis of isopentenyl diphosphate (IPP) and its isomer dimethylallyl diphosphate (DMAPP), the precursors of all isoprenoid compounds. The 4-diphosphocytidyl-2-C-methyl-D-erythritol synthase (IspD; EC 2.7.7.60) is the key enzyme of the MEP pathway. It is also of interest as a new chemotherapeutic target, as the enzyme is absent in mammals and ispD is an essential gene for growth. A high-throughput screening method was therefore developed to identify compounds that inhibit IspD. This process was applied to identify a lead compound, domiphen bromide (DMB), that may effectively inhibit IspD. The inhibitory action of DMB was confirmed by over-expressing or down-regulating IspD in Mycobacterium smegmatis (M. smegmatis), demonstrating that DMB inhibit M. smegmatis growth additionally through an IspD-independent pathway. This also led to higher levels of growth inhibition when combined with IspD knockdown. This novel IspD inhibitor was also reported to exhibit antimycobacterial activity in vitro, an effect that likely occurs as a result of perturbation of cell wall biosynthesis.
Sigma-Aldrich
Domiphen bromide, 97%