MilliporeSigma
  • An efficient one-pot synthesis of pyrano[3,2-c]quinolin-2,5-dione derivatives catalyzed by L-proline.
An efficient one-pot synthesis of pyrano[3,2-c]quinolin-2,5-dione derivatives catalyzed by L-proline.
Molecules (Basel, Switzerland) (2012-11-24)
Songlei Zhu, Jing Wang, Zhou Xu, Jie Li
ABSTRACT
A series of 4-aryl-6-methyl-3,4-dihydro-2H-pyrano[3,2-c]quinolin-2,5(6H)-diones were synthesized via the three-component reactions of aromatic aldehydes, 4-hydroxy-1-methylquinolin-2(1H)-one, and Meldrum's acid catalyzed by L-proline. The structures of the products were identified by spectroscopic analysis. A mechanism for this three-component reaction catalyzed by L-proline was proposed.
Sigma-Aldrich
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%