MilliporeSigma
  • Species-specific lipophilicity of thyroid hormones and their precursors in view of their membrane transport properties.
Species-specific lipophilicity of thyroid hormones and their precursors in view of their membrane transport properties.
Journal of pharmaceutical and biomedical analysis (2013-01-10)
Gergő Tóth, Károly Mazák, Sándor Hosztafi, József Kökösi, Béla Noszál
ABSTRACT
A total of 30 species-specific partition coefficients of three thyroid hormones (thyroxine, liothyronine, reverse liothyronine) and their two biological precursors (monoiodotyrosine, diiodotyrosine) are presented. The molecules were studied using combined methods of microspeciation and lipophilicity. Microspeciation was carried out by (1)H NMR-pH and UV-pH titration techniques on the title compounds and their auxiliary derivatives of reduced complexity. Partition of some of the individual microspecies was mimicked by model compounds of the closest possible similarity, then correction factors were determined and introduced. Our data show that the iodinated aromatic ring system is the definitive structural element that fundamentally determines the lipophilicity of thyroid hormones, whereas the protonation state of the aliphatic part plays a role of secondary importance. On the other hand, the lipophilicity of the precursors is highly influenced by the protonation state due to the relative lack of overwhelmingly lipophilic moieties. The different logp values of the positional isomers liothyronine and reverse liothyronine represent the importance of steric and electronic factors in lipophilicity. Our investigations provided clear indication that overall partition, the best membrane transport - predicting physico-chemical parameter depends collectively on the site-specific basicity and species-specific partition coefficient. At physiological pH these biomolecules are strongly amphipathic due to the lipophilic aromatic rings and hydrophilic amino acid side chains which can well be the reason why thyroid hormones cannot cross membranes by passive diffusion and they are constituents of biological membranes. The lipophilicity profile of thyroid hormones and their precursors are calculated and depicted in terms of species-specific lipophilicities over the entire pH range.
Product Description
Supelco
3,3′,5′-Triiodo-L-thyronine (Reverse T3) solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®