MilliporeSigma
  • Tris acetylacetonate aluminium(III) induces osmotic fragility and acanthocyte formation in suspended erythrocytes.
Tris acetylacetonate aluminium(III) induces osmotic fragility and acanthocyte formation in suspended erythrocytes.
Toxicology letters (1989-01-01)
P Zatta, M Perazzolo, B Corain
ABSTRACT
Tris acetylacetonate aluminium(III) (Al(acac)3), dissolved in water, is effective in producing osmotic fragility in suspended erythrocytes in the concentration range of 0.034-0.34 mmol/l. Water solutions of Tris maltolate aluminium(III) (Al(malt)3) and aluminium lactate (Al(lac)3) are also effective but the dose-response behavior is less pronounced. Moreover, only Al(acac)3 induces a prominent generation of acanthocytes. The stronger effects of Al(acac)3 on membrane stability are attributed to the greater solubility of this complex in the cell membrane.
Sigma-Aldrich
Aluminum acetylacetonate, 98%
Sigma-Aldrich
Aluminum acetylacetonate, purified by sublimation, 99.999% trace metals basis