MilliporeSigma
The kininases of Bothrops jararaca plasma.
Acta physiologica latino americana (1980-01-01)
A A Lavras, M Fichman, E Hiraichi, T Tobo, M A Boucault
ABSTRACT
Evidence is presented to suggest that kininase activity of Bothrops jararaca plasma is due to the presence of at least three distinct enzymes: a carboxypeptidase B type enzyme, similar to that found in human plasma in that its activity is enhanced by Co2+ (1 X 10(-4) M); a carboxypeptidase B type enzyme whose activity is unaffected by Co2+, and an enzyme which cleaves bradykinin to liberate Phe-Arg as the major peptide fragment formed. The latter enzyme is responsible for the major kininase activity of this snake plasma and is identified as a dipeptide hydrolase.
Sigma-Aldrich
Hippuryl-Lys