MilliporeSigma
Kinetics of bupivacaine after levcromakalim treatment in mice.
The Journal of pharmacy and pharmacology (1997-03-01)
M Gantenbein, L Attolini, B Bruguerolle
ABSTRACT
Previous workers have reported that 0.01 mg kg-1 of levcromakalim injected intraperitoneally did not modify bupivacaine-induced neurotoxicity but increased the duration of action of bupivacaine. This study was designed to document possible changes in the pharmacokinetic behaviour of bupivacaine and its main metabolite, N-desbutylbupivacaine in mice after a single 0.01 mg kg-1 intraperitoneal injection of levcromakalim. The kinetic parameters of bupivacaine were determined after a single 20 mg kg-1 intraperitoneal injection of bupivacaine in controls and in levcromakalim-treated mice. It was found that levcromakalim did not change any kinetic parameters of bupivacaine or of its main metabolite, N-desbutylbupivacaine. The previously reported findings of the influence of the low dose (0.01 mg kg-1) of levcromakalim on bupivacaine-induced toxicity agree well with the lack of influence of 0.01 mg kg-1 of levcromakalim on bupivacaine and N-desbutylbupivacaine pharmacokinetics, although the reported increase in the duration of action of bupivacaine after levcromakalim treatment can hardly be explained by the pharmacokinetics of bupivacaine when associated with levcromakalim. We suggest that levcromakalim might interfere directly with ion-channel block caused by bupivacaine by altering conduction properties or indirectly by enhancing bupivacaine-induced voltage and time-dependent sodium-channel block.
Product Description
Mepivacaine impurity B, European Pharmacopoeia (EP) Reference Standard