MilliporeSigma
  • Analytical derivatizations of volatile and hydrophilic carbonyls from aqueous matrix onto a solid phase of a polystyrene-divinylbenzene macroreticular resin.
Analytical derivatizations of volatile and hydrophilic carbonyls from aqueous matrix onto a solid phase of a polystyrene-divinylbenzene macroreticular resin.
Journal of chromatography. B, Biomedical sciences and applications (1997-07-04)
S M Breckenridge, X Yin, J M Rosenfeld, Y H Yu
ABSTRACT
Extraction and derivatization of carbonyls to benzyloximes, pentafluorobenzyloximes or 2,4-dinitrophenylhydrazones is simplified and reaction times are substantially reduced by simultaneous sorption and derivatization from aqueous solution onto a solid phase. In this reaction a macroreticular polystyrene-divinylbenzene resin acts as a sorbent and catalyst to allow simultaneous extraction and derivatization of hydrophilic and lipophilic aldehydes and ketones from simple as well as complex matrices including plasma. Conversion to the 2,4-dinitrophenylhydrazones or pentafluorobenzyloximation at ambient temperature requires 10 and 20 min, respectively. These reaction conditions correspond to at least a 6-fold reduction in reaction times for derivatization of the reactive aldelhydes and a 36-72-fold reduction for preparation of derivatives for the slower reacting ketones.
Sigma-Aldrich
O-Benzylhydroxylamine hydrochloride, 99%