MilliporeSigma
  • Detection of hemolysin variants of Shiga toxin-producing Escherichia coli by PCR and culture on vancomycin-cefixime-cefsulodin blood agar.
Detection of hemolysin variants of Shiga toxin-producing Escherichia coli by PCR and culture on vancomycin-cefixime-cefsulodin blood agar.
Applied and environmental microbiology (1998-07-02)
A Lehmacher, H Meier, S Aleksic, J Bockemühl
ABSTRACT
The presence of a hemolysin-encoding gene, elyA or hlyA, from Shiga toxin-producing Escherichia coli (STEC) was detected by PCR in each of 95 strains tested. PCR products of elyA from human STEC isolates of serovars frequently detected in Germany, such as O157:H-, O103:H2, O103:H-, O26:H11, and O26:H-, showed nucleotide sequences identical to previously reported ones for O157:H7 and O111:H- strains. Compared to them, four elyA amplicons derived from human isolates of rare STEC serovars showed identity of about 98% but lacked an AluI restriction site. However, the nucleotide sequence of an amplicon derived from a porcine O138:K81:H- STEC strain was identical to the corresponding region of hlyA, encoding alpha-hemolysin, from E. coli. This hlyA amplicon showed 68% identity with the nucleotide sequence of the corresponding elyA fragment. It differed from the elyA PCR product in restriction fragments generated by AluI, EcoRI, and MluI. Of the 95 representative STEC strains, 88 produced hemolysin on blood agar supplemented with vancomycin (30 mg/liter), cefixime (20 micrograms/liter), and cefsulodin (3 mg/liter) (BVCC). The lowest added numbers of two to six STEC CFU per g of stool or per ml of raw milk were detectable on BVCC plates after seeding of the preenrichment broth, modified tryptic soy broth (mTSB) supplemented with novobiocin (10 mg/liter), with 16 STEC strains. These strains represented the seven prevailing serovars diagnosed from German patients. However, with ground-beef samples, PCR was essential to identify the lowest added numbers of two to six STEC CFU among colonies of hemolyzing Enterobacteriaceae, such as Serratia spp. and alpha-hemolysin-producing E. coli. We conclude that preenrichment of stool and food samples in mTSB for 6 h followed by overnight culturing on BVCC is a simple method for the isolation and presumptive identification of STEC.
Sigma-Aldrich
Cefsulodin sodium salt hydrate, third-generation cephalosporin antibiotic