MilliporeSigma
  • Colloidal synthesis of inorganic fullerene nanoparticles and hollow spheres of titanium disulfide.
Colloidal synthesis of inorganic fullerene nanoparticles and hollow spheres of titanium disulfide.
Chemical communications (Cambridge, England) (2010-09-21)
Sujay Prabakar, Sean Collins, Bryan Northover, Richard D Tilley
ABSTRACT
The synthesis of inorganic fullerene (IF) nanoparticles and IF hollow spheres of titanium disulfide by a simple colloidal route is reported. The injection temperature of the titanium precursor into the solvent mixture was found to be important in controlling the morphology.
Sigma-Aldrich
Titanium(IV) sulfide, powder, -200 mesh, 99.9%