MilliporeSigma
  • Addition of TMSCN to chiral ketimines derived from isatin. Synthesis of an oxindole-based peptidomimetic and a bioactive spirohydantoin.
Addition of TMSCN to chiral ketimines derived from isatin. Synthesis of an oxindole-based peptidomimetic and a bioactive spirohydantoin.
Organic & biomolecular chemistry (2011-06-21)
Alessandro Sacchetti, Alessandra Silvani, Francesco G Gatti, Giordano Lesma, Tullio Pilati, Beatrice Trucchi
ABSTRACT
We investigated the Strecker-type reaction of isatin derived chiral ketimines with TMSCN in the presence of a Lewis acid. The desired α-amino nitriles have been obtained in good yields with moderate diastereoselectivity. Further elaboration of the cyanide group allowed the preparation of a new oxindole-based peptidomimetic and a pharmaceutically relevant spirohydantoin.
Sigma-Aldrich
Trimethylsilyl cyanide, technical, ≥95% (GC)
Sigma-Aldrich
Trimethylsilyl cyanide, 98%