MilliporeSigma
  • Urotricha psenneri n. sp. and Amphileptus piger (Vuxanovici, 1962) n. comb., two planktonic ciliates (Protozoa, Ciliophora) from an oligotrophic lake in Austria.
Urotricha psenneri n. sp. and Amphileptus piger (Vuxanovici, 1962) n. comb., two planktonic ciliates (Protozoa, Ciliophora) from an oligotrophic lake in Austria.
The Journal of eukaryotic microbiology (2005-01-26)
Bettina Sonntag, Wilhelm Foissner
ABSTRACT
Two euplanktonic ciliates, Urotricha psenneri n. sp. (Prostomatida) and Amphileptus piger (Vuxanovici, 1962) n. comb. (Pleurostomatida), were discovered in the surface plankton of the oligotrophic Lake Traunsee in Austria. Their morphology and infraciliature were studied in live cells as well as in specimens impregnated with protargol and silver nitrate. Urotricha psenneri is a small urotrichid, less than 50 microm length and with a single caudal cilium. It is unique in having (i) a massive oral basket projecting as a conspicuous bulge with cylindrical microfibrillar annulus and (ii) a curved brosse row 1 in the broad, barren circumoral area. Amphileptus piger (Vuxanovici, 1962) is about 55 x 13 microm in vivo, has two macronuclear nodules with a single micronucleus in between in the posterior body half, has a single contractile vacuole with a terminal excretory pore, and few, but thick and thus highly conspicuous extrusomes. The amphileptid ciliary pattern (spica) is difficult to recognise due to the widely spaced basal bodies.
Sigma-Aldrich
Silver proteinate, ~8% Ag basis