MilliporeSigma
[Treatment of acute rhinitis in neonates and infants].
Vestnik otorinolaringologii (2005-02-09)
V S Minasian, M G Bondarenko
ABSTRACT
The authors review current views on the treatment of acute rhinitis (AR) in infants and present their experience in AR treatment of 30 infants aged from 3 days to 1 year. Combined treatment with Physiomer baby quickly eradicated inflammation. The drug eliminates crusts and nasal discharge allowing application of other medication on a clear surface of nasal mucosa. As a result, medication becomes more affective and accelerates recovery. A convenient tip is well adapted for the nostrils. Administration of medicine by spray provides optimal lavage of the nasal cavity.
Sigma-Aldrich
Silver proteinate, ~8% Ag basis