MilliporeSigma
  • (1)H and (13)C NMR studies of asymmetrically substituted bis- and tris-Tröger's bases.
(1)H and (13)C NMR studies of asymmetrically substituted bis- and tris-Tröger's bases.
Magnetic resonance in chemistry : MRC (2005-06-30)
J Elguero, A Fruchier, T Mas, C Pardo
ABSTRACT
The (1)H and (13)C NMR spectra of two stereoisomeric bis-Tröger's bases and four stereoisomeric tris-Tröger's bases asymmetrically substituted on the external aromatic rings were recorded and the corresponding signals assigned. The relative configuration of the stereogenic units has been unequivocally determined on the basis of homoallylic couplings and NOE experiments.
Product Description
Troger′s Base, 98%