MilliporeSigma
  • Synthesis and SAR of 2-phenyl-1-sulfonylaminocyclopropane carboxylates as ADAMTS-5 (Aggrecanase-2) inhibitors.
Synthesis and SAR of 2-phenyl-1-sulfonylaminocyclopropane carboxylates as ADAMTS-5 (Aggrecanase-2) inhibitors.
Bioorganic & medicinal chemistry letters (2009-09-22)
Makoto Shiozaki, Hiroto Imai, Katsuya Maeda, Tomoya Miura, Katsutaka Yasue, Akira Suma, Masahiro Yokota, Yosuke Ogoshi, Julia Haas, Andrew M Fryer, Ellen R Laird, Nicole M Littmann, Steven W Andrews, John A Josey, Takayuki Mimura, Yuichi Shinozaki, Hiromi Yoshiuchi, Takashi Inaba
ABSTRACT
A series of 1-sulfonylaminocyclopropanecarboxylates was synthesized as ADAMTS-5 (Aggrecanase-2) inhibitors. After an intensive investigation of the central cyclopropane core including its absolute stereochemistry and substituents, we found compound 22 with an Agg-2 IC50=7.4 nM, the most potent ADAMTS-5 inhibitor reported so far.
Sigma-Aldrich
Cyclopropanecarboxylic acid, 95%