MilliporeSigma
  • Hypoglycemic and antioxidative effects of hydroxyethyl methylcellulose in mice fed with high fat diet.
Hypoglycemic and antioxidative effects of hydroxyethyl methylcellulose in mice fed with high fat diet.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-03-06)
Su Jeong Ban, Catherine W Rico, In Chul Um, Mi Young Kang
ABSTRACT
The effect of hydroxyethyl methylcellulose (HEMC) with different viscosities on the glucose metabolism and antioxidative defense system in high fat-fed mice was investigated. The mice were randomly divided into five dietary groups: normal control diet (NC), high fat diet (HF), and high fat diet supplemented with high viscosity (HF-HV), moderate viscosity (HF-MV), and low viscosity (HF-LV) HEMC fibers. After 6 weeks, the HF group showed a marked increase in body weight gain, body fat, blood glucose concentration, insulin level, and erythrocyte lipid peroxidation rate relative to the NC group. However, supplementation of HEMC in the diet suppressed these high fat-induced hyperglycemia and oxidative stress through enhancement of the activities of hepatic glucokinase and antioxidant enzymes. The hypoglycemic and antioxidative effects increased with increased viscosity of the HEMC consumed. These results illustrate that HEMC with high viscosity may be useful in the management of high fat diet-induced hyperglycemia and oxidative stress.
Sigma-Aldrich
Tylose® MH 1000, viscosity 600-1500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Tylose® MH 300, viscosity 150-450 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl 2-hydroxyethyl cellulose