MilliporeSigma
  • Endogenous regeneration of damaged retinal pigment epithelium following low dose sodium iodate administration: an insight into the role of glial cells in retinal repair.
Endogenous regeneration of damaged retinal pigment epithelium following low dose sodium iodate administration: an insight into the role of glial cells in retinal repair.
Experimental eye research (2013-04-30)
Anna Machalińska, Miłosz Piotr Kawa, Ewa Pius-Sadowska, Dorota Rogińska, Patrycja Kłos, Bartłomiej Baumert, Barbara Wiszniewska, Bogusław Machaliński
ABSTRACT
The retinal pigment epithelium (RPE) has been reported to demonstrate feasible self-regenerative potential under specific conditions. However, the precise underlying mechanisms involved in this process are still elusive. Here, we performed a sequential morphological, molecular, and functional analysis of retinal injury and subsequent tissue regeneration after intravenous administration of a low dose of sodium iodate (15 mg/kg) in mice over long-term observation, up to 3 months post-injury. To assess the kinetics of the injury/recovery process, the electroretinography (ERG) responses were correlated with ongoing alterations in retinal structure and the global gene expression profile of injured retinas using genome-wide RNA microarray technology, western blotting and immunohistochemical analyses. We observed considerable improvement in the rod cell-mediated ERG response, which was accompanied by the regeneration of RPE within the injury site by the 3rd month post-injury. Our results confirm that the repairing mechanisms within injured retinas involve a significant glial cell reaction marked by glial cell proliferation, migration from their original location toward the injury site, followed by a significant overproduction of NTs such as BDNF, GDNF and NT-3. The global gene expression analysis revealed that initially up-regulated genes associated with cell death, apoptosis, acute response to stress pathways underwent considerable down-regulation in the late post-injury period. Accordingly, the genes implicated in nervous tissue remodeling and neuron development, the regulation of synaptic transmission and the establishment of localization were substantially induced by the 3rd month. Collectively, our observations support the view that Müller glial cells might well play an active role not only in retinal cell reorganization following injury but potentially also in RPE regeneration, which appears to be the key event in retinal reparative process. Furthermore, we provided novel compelling evidence of the crucial role of neurotrophins in the pathophysiology of retinal repair and identified the signaling pathways that are activated during this process.
Sigma-Aldrich
Sodium iodate, ≥99%
Sigma-Aldrich
Sodium iodate, puriss. p.a., ≥99.5% (RT)