MilliporeSigma
  • Activity of WY-49605 compared with those of amoxicillin, amoxicillin-clavulanate, imipenem, ciprofloxacin, cefaclor, cefpodoxime, cefuroxime, clindamycin, and metronidazole against 384 anaerobic bacteria.
Activity of WY-49605 compared with those of amoxicillin, amoxicillin-clavulanate, imipenem, ciprofloxacin, cefaclor, cefpodoxime, cefuroxime, clindamycin, and metronidazole against 384 anaerobic bacteria.
Antimicrobial agents and chemotherapy (1994-11-01)
S K Spangler, M R Jacobs, P C Appelbaum
ABSTRACT
The National Committee for Clinical Laboratory Standards agar dilution method was used to compare the in vitro activity of WY-49605 (also called SUN/SY 5555 and ALP-201), a new broad-spectrum oral penem, to those of amoxicillin, amoxicillin-clavulanate, imipenem, ciprofloxacin, cefaclor, cefpodoxime, cefuroxime, clindamycin, and metronidazole against 384 clinically isolated anaerobes. These anaerobic organisms included 90 strains from the Bacteroides fragilis group, 87 Prevotella and Porphyromonas strains, non-B. fragilis group Bacteroides strains, 56 fusobacteria, 55 peptostreptococci, 49 gram-positive non-spore-forming rods, and 47 clostridia. Overall, WY-49605 had an MIC range of 0.015 to 8.0 micrograms/ml, an MIC at which 50% of the isolates are inhibited (MIC50) of 0.25 microgram/ml, and an MIC at which 90% of the isolates are inhibited (MIC90) of 2.0 micrograms/ml. Good activity against all anaerobe groups was observed, except for Clostridium difficile and lactobacilli (MIC50s of 4.0 and 2.0 micrograms/ml, respectively, and MIC90s of 8.0 and 2.0 micrograms/ml, respectively). Imipenem had an MIC50 of 0.03 microgram/ml and an MIC90 of 0.25 microgram/ml. Ciprofloxacin was much less active (MIC50 of 2.0 micrograms/ml and MIC90 of 16.0 micrograms/ml). By comparison, all oral beta-lactams were less active than WY-49605, with susceptibilities as follows: amoxicillin MIC50 of 8.0 micrograms/ml and MIC90 of > 256.0 micrograms/ml), amoxicillin-clavulanate MIC50 of 1.0 microgram/ml and MIC90 of 8.0 micrograms/ml, cefaclor MIC50 of 8.0 micrograms/ml and MIC90 of > 32.0 micrograms/ml, cefpodoxime MIC50 of 4.0 micrograms/ml and MIC90 of > 32.0 micrograms/ml, and cefuroxime MIC50 of 4.0 micrograms/ml and MIC90 of > 32.0 micrograms/ml. Clindamycin was active against all groups except some members of the B. fragilis group, Fusobacterium varium, and some clostridia ( overall MIC50 of 0.5 micrograms/ml and overall MIC90 of 8.0 micrograms/ml). Metronidazole was active (MIC of less than or equal to 4.0 micrograms/ml) against all gram-negative anaerobic rods, but most gram-positive non-spore-forming rods, some peptostreptococci, and some clostridia were less susceptible. To date, WY-49605 is the most active oral beta-lactam against anaerobes: these results suggest clinical evaluation for clinical indications suitable for oral therapy.
Sigma-Aldrich
Faropenem sodium hydrate, ≥98% (HPLC)