MilliporeSigma
Presence of an O-glycosidically linked hexasaccharide in fetuin.
The Journal of biological chemistry (1987-11-25)
A S Edge, R G Spiro
ABSTRACT
Examination by gel filtration, thin layer and anion exchange chromatography of the O-linked carbohydrate units released from fetuin by alkaline borohydride treatment indicated the presence in this glycoprotein of an acidic glucosamine-containing hexasaccharide in addition to the previously described tetra- and trisaccharides. The structure of the hexasaccharide was determined to be NeuAc alpha 2----3Gal beta 1----3[NeuAc alpha 2----3Gal beta 1----4GlNAc beta 1----6]GalNAc, on the basis of exoglycosidase digestion, periodate oxidation, and methylation analysis as well as hydrazine-nitrous acid fragmentation. The latter procedure when carried out on the reduced asialohexasaccharide yielded Gal----2-deoxygalactitol and Gal----anhydromannose which were shown to be derived, respectively, from Gal----N-acetylgalactosaminitol and Gal----GlcNAc sequences. Reductive amination of the Gal----anhydromannose disaccharide with [14C] methylamine permitted identification of its linkage as 1----4. While Diplococcus pneumoniae endo-alpha-DN-acetylgalactosaminidase acting on asialofetuin released the sialic acid-free tetra- and trisaccharides (Gal beta 1----3GalNAc), this enzyme did not cleave the peptide attachment of the asialohexasaccharide (Gal beta 1----3 [Gal beta 1----4GlcNAc beta 1----6] GalNAc). The number of O-linked hexa-, tetra-, and trisaccharides per fetuin molecule was determined to be 0.2, 0.7, and 2.1, respectively, on the basis of galactosaminitol analyses. The absence of O-linked N-acetylglucosamine-containing tetra- or pentasaccharides in fetuin suggest that the attachment of this sugar is a rate-limiting step; furthermore, the limited occurrence of the hexasaccharide may indicate that the addition of sialic acid to Gal beta 1----3GalNAc to form the NeuAc alpha 2----3Gal linkage precludes action of the GlcNAc transferase to form the branch point on the GalNAc residue.
Sigma-Aldrich
3′-N-Acetylneuraminyl-N-acetyllactosamine sodium salt, ≥95%