MilliporeSigma
  • Cetirizine as pH-dependent cross-reactant in a carbamazepine-specific immunoassay.
Cetirizine as pH-dependent cross-reactant in a carbamazepine-specific immunoassay.
The Analyst (2011-02-19)
Arnold Bahlmann, Jana Falkenhagen, Michael G Weller, Ulrich Panne, Rudolf J Schneider
ABSTRACT
High performance liquid chromatography (HPLC) was hyphenated with a previously reported carbamazepine-specific enzyme-linked immunosorbent assay (ELISA) as a screening approach to water analysis in order to identify possible interferences from transformation products. Treated wastewater was analysed and three substances were recognized by the antibody besides carbamazepine: the metabolites 10,11-dihydro-10,11-epoxycarbamazepine and 2-hydroxycarbamazepine plus the structurally not obviously related antihistamine cetirizine. The molar cross-reactivity against cetirizine was found to be pH-dependent and assessed to be 400% at pH 4.5 and 22% at pH 10.5. Performing the ELISA at pH 10.5 greatly improved the accuracy when carbamazepine was determined in surface and wastewater samples.
Sigma-Aldrich
4-Chlorobenzhydrol, 98%