MilliporeSigma
  • Synthesis of tetrahydropyrimidin-2-ones via FeCl3 catalyzed one-pot domino reaction of amines, methyl propiolate, aromatic aldehydes, and urea.
Synthesis of tetrahydropyrimidin-2-ones via FeCl3 catalyzed one-pot domino reaction of amines, methyl propiolate, aromatic aldehydes, and urea.
Molecular diversity (2014-01-31)
Li-Li Zhang, Jing Sun, Chao-Guo Yan
ABSTRACT
Polysubstituted 3-arylaminoacrylate and tetrahydropyrimidin-2-one derivatives could be selectively produced from the one-pot domino reaction of arylamines, methyl propiolate, aromatic aldehydes, and urea in ethanol in the presence of FeCl3 as catalyst. Under similar reactions secondary amines such as morpholine and piperidine predominately afford tetrahydropyrimidin-2-one derivatives in good yields.
Sigma-Aldrich
Methyl propiolate, 99%