MilliporeSigma
Use of clomiphene citrate in infertile women: a committee opinion.
Fertility and sterility (2013-07-03)
ABSTRACT
This committee opinion describes the use of clomiphene citrate, including indications, use, monitoring, and side effects. There is also a discussion of adjuvants and alternatives to clomiphene citrate therapy.
Sigma-Aldrich
Enclomiphene hydrochloride, ≥96% (HPLC)