MilliporeSigma
  • Total synthesis of (-)-Penifulvin A, an insecticide with a dioxafenestrane skeleton.
Total synthesis of (-)-Penifulvin A, an insecticide with a dioxafenestrane skeleton.
Journal of the American Chemical Society (2009-01-15)
Tanja Gaich, Johann Mulzer
ABSTRACT
Herein we report the first total synthesis of Penifulvin A, a sesquiterpenoid with a novel dioxa-fenestrane structure. Penifulvin A is a potent insecticide against the fall armyworm Spodoptera frugiperda which causes enormous damage in the US by consuming foliage of a variety of field crops. A five-step racemic and an eight-step enantioselective route to the natural product and the determination of its absolute configuration are described. The key step involves a meta-photocycloaddition, giving rapid access to the carbon skeleton of penifulvin A in a stereoselective fashion. Finally an oxidation cascade leads to the natural product. The synthetic route is free from protecting groups, scalable, and flexible so that a variety of analogues, among them penifulvins B-E, should be available for performing SAR tests in the insecticidal role.
Sigma-Aldrich
5-Bromo-2-methyl-2-pentene, 97%