MilliporeSigma
  • Heterogeneity of tonsillar subepithelial B lymphocytes, the splenic marginal zone equivalents.
Heterogeneity of tonsillar subepithelial B lymphocytes, the splenic marginal zone equivalents.
Journal of immunology (Baltimore, Md. : 1950) (2000-05-23)
M Dono, S Zupo, N Leanza, G Melioli, M Fogli, A Melagrana, N Chiorazzi, M Ferrarini
ABSTRACT
The VH4 genes expressed by both resting and in vivo-activated subepithelial (SE) B cells from human tonsils were studied. Resting SE B cells were subdivided according to the presence (IgDlow) or absence (IgM-only) of surface IgD. CD27 was abundant on activated SE B cells and low on resting IgM-only B cells. Resting IgDlow SE B cells could be subdivided into CD27low and CD27high cell fractions. Resting IgDlow SE B cells displayed VH4 genes with a substantial number of mutations (13/29 of the molecular clones were mutated), whereas 25/26 of the clones from resting IgM-only SE B cells were unmutated. Moreover, mutated VH4 genes were detected mainly within the CD27high cell fraction of the IgDlow SE B cells. Several identical unmutated VH4DJH sequences (11/32) were found in different molecular clones from resting IgM-only SE B cells, suggesting local cellular expansion. Both unmutated (14/25) and mutated (11/25) sequences were found in mu transcripts of activated SE B cells. Extensive mutation was observed in the gamma transcripts of activated SE B cells. Therefore, SE B cells are heterogeneous, being comprised of B cells with mutated Ig VH4 genes, that are Ag-experienced B cells, and a subset of B cells with unmutated VH4 genes that are either virgin cells or cells driven by Ags that did not induce or select for V gene mutations.
Sigma-Aldrich
CD38 (SP149) Rabbit Monoclonal Antibody