MilliporeSigma
  • Relationship between polymorphisms in beta -2 adrenergic receptor gene and ischemic stroke in North Indian Population: a hospital based case control study.
Relationship between polymorphisms in beta -2 adrenergic receptor gene and ischemic stroke in North Indian Population: a hospital based case control study.
BMC research notes (2014-06-27)
Amit Kumar, Manjari Tripathi, Madakasira Vasantha Padma Srivastava, Subbiah Vivekanandhan, Kameshwar Prasad
ABSTRACT
Stroke is a multi-factorial disease and influenced by both genetic and environmental factors. The purpose of the present case control study was to check the relationship between beta-2 adrenergic receptor (ADRB2) polymorphism and ischemic stroke in North Indian Population. In a hospital based case control study, patients with ischemic stroke and control subjects from outpatient department and neurology ward of All India Institute of Medical Sciences New Delhi. Genotyping was performed by using Polymerase chain reaction-Restriction fragment length polymorphism. Frequency distributions of genotypes and alleles were compared between cases and controls using multivariate logistic regression. In this study, 224 patients and 224 age-and sex-matched control subjects were recruited. Mean age of cases and controls were 53.9 ± 13.4 and 53.6 ± 12.9 years respectively. Multivariate logistic regression analysis showed an independent association between Gln27Glu polymorphism and large vessel stroke (LVD) under a recessive model of inheritance (OR 3.9; 95% CI 1.3 to 11). An age-stratified analysis, suggested independent association between Gln27Glu polymorphism and ischemic stroke, large vessel disease and small vessel disease stroke who had onset of disease at an older age. The findings of the present study suggest that Gln27Glu polymorphism of the ADRB2 gene may confer higher risk of large vessel disease stroke in a North Indian population. Prospective studies with larger sample size are required for independent validation.