MilliporeSigma
  • Enzyme reactor for urinary acylcarnitines assay by reversed-phase high-performance liquid chromatography.
Enzyme reactor for urinary acylcarnitines assay by reversed-phase high-performance liquid chromatography.
Clinica chimica acta; international journal of clinical chemistry (1993-07-16)
K Matsumoto, M Takahashi, N Takiyama, H Misaki, N Matsuo, S Murano, H Yuki
ABSTRACT
An immobilized enzyme reactor, made up acylcarnitine hydrolase, carnitine dehydrogenase and diaphorase in sequence, was developed for the sensitive and selective determination of urinary free and individual acylcarnitines by a reversed-phase high-performance liquid chromatography. A 100-microliter urine sample was directly injected onto the TSKgel ODS 80Ts column and eluted by a step-gradient procedure. The eluent was mixed with the substrate solution of beta-NAD+ (1.0 mmol/l), resazurin (25 mumol/l) and Tris acetate (0.2 mol/l, pH 9.0). The mixture was passed through the immobilized enzyme reactor at 40 degrees C. Acylcarnitines were hydrolyzed and the converted to rezorufin which was measured by monitoring the fluorescence intensity at lambda EX = 560 nm and lambda EM = 580 nm. Free, acetyl-, glutaryl-, propionyl-, butyryl-, isobutyryl-, valeryl- and isovalerylcarnitine were determined within 55 min with detection limits (< 1 mumol/l) and within-run and day-to-day imprecision (C.V. < 6%). Free, acetyl- and isobutyrylcarnitine were found in normal urine. On the other hand, propionylcarnitine was detected in the urine of children with propionic aciduria and methylmalonic aciduria and multiple acylcarnitines were found in the urine of children with glutaric aciduria (type II).
Sigma-Aldrich
Isobutyryl-L-carnitine, ≥97.0% (HPLC)