MilliporeSigma
  • Synthesis and antimicrobial activity of new 4-heteroarylamino coumarin derivatives containing nitrogen and sulfur as heteroatoms.
Synthesis and antimicrobial activity of new 4-heteroarylamino coumarin derivatives containing nitrogen and sulfur as heteroatoms.
Molecules (Basel, Switzerland) (2010-04-30)
Biljana R Dekić, Niko S Radulović, Vidoslav S Dekić, Rastko D Vukićević, Radosav M Palić
ABSTRACT
Synthesis, spectral analysis and bioactivity of new coumarin derivatives are described in this paper. Eight new coumarin derivatives were synthesized in moderate to good yields by condensation of 4-chloro-3-nitrocoumarin and the corresponding heteroarylamine. The synthesized compounds were tested for their in vitro antimicrobial activity, in a standard disk diffusion assay, against thirteen strains of bacteria and three fungal strains. They have shown a wide range of activity - from one completely inactive compound to medium active ones.
Sigma-Aldrich
4-Hydroxy-3-nitrocoumarin, 98%