MilliporeSigma
  • Steric stabilization of liposomes by pH-responsive N-isopropylacrylamide copolymer.
Steric stabilization of liposomes by pH-responsive N-isopropylacrylamide copolymer.
Journal of pharmaceutical sciences (2002-07-13)
Emmanuelle Roux, Romain Stomp, Suzanne Giasson, Michel Pézolet, Pierre Moreau, Jean-Christophe Leroux
ABSTRACT
The aim of this study was to characterize a pH-sensitive liposome formulation bearing a terminally alkylated N-isopropylacrylamide (NIPAM) copolymer with regard to its pH responsiveness, surface properties, and pharmacokinetics. The interacting forces between two lipid bilayers bearing the anchored NIPAM copolymer were measured with a surface force apparatus. The pH-triggered content release was evaluated in buffer before and after incubation in human serum. The pharmacokinetics was determined in rats following the intravenous injection of 67Ga-loaded liposomes with or without the polymer coating. The force measurements between lipid bilayers showed that NIPAM copolymers provide a steric barrier that was dependent on pH. The pH-sensitive liposomes maintained their pH sensitivity after incubation in serum. In vivo, the polymer-coated liposomes exhibited a prolonged circulation time in rats, with an area under the blood concentration-time curve that is 1.6-fold higher than the control formulation. This study showed that liposomes can be rendered pH sensitive by anchoring a terminally alkylated NIPAM copolymer at their surface. At neutral pH, the polymer provides a steric barrier that increases the liposome circulation time in vivo.
Sigma-Aldrich
Dioctadecylamine, ≥99.0% (NT)