MilliporeSigma
  • Possible intermediates of Cu(phen)-catalyzed C-O cross-coupling of phenol with an aryl bromide by in situ ESI-MS and EPR studies.
Possible intermediates of Cu(phen)-catalyzed C-O cross-coupling of phenol with an aryl bromide by in situ ESI-MS and EPR studies.
Dalton transactions (Cambridge, England : 2003) (2014-06-18)
Hong-Jie Chen, I-Jui Hsu, Mei-Chun Tseng, Shin-Guang Shyu
ABSTRACT
The C-O coupling reaction between 2,4-dimethylphenol and 4-bromotoluene catalyzed by the CuI/K2CO3/phen system can be inhibited by the radical scavenger cumene. Complexes [Cu(i)(phen)(1-(2,4-dimethylphenoxy)-4-methylbenzene)](+) (denoted as A), {H[Cu(i)(phen)(2,4-dimethylphenoxy)]}(+) and [Cu(i)(2,4-dimethylphenoxy)2](-) (denoted as B) were observed by in situ electrospray ionization mass spectrometry (ESI-MS) analysis of the copper(i)-catalyzed C-O coupling reaction under the catalytic reaction conditions indicating that they could be intermediates in the reaction. The in situ EPR study of the reaction solution detected the Cu(ii) species with a fitted g value of 2.188. A catalytic cycle with a single electron transfer (SET) step was proposed based on these observations.
Sigma-Aldrich
4-Bromotoluene, 98%