MilliporeSigma
  • Supramolecular phthalocyanine dimers based on the secondary dialkylammonium cation/dibenzo-24-crown-8 recognition motif
Supramolecular phthalocyanine dimers based on the secondary dialkylammonium cation/dibenzo-24-crown-8 recognition motif
Organic letters (2000-05-11)
Martinez-Diaz, Rodriguez-Morgade, Feiters, van Kan PJ, Nolte, Stoddart, Torres
ABSTRACT
[formula: see text] New unsymmetrically substituted DB24C8-phthalocyanines, which are able to form complexes with suitable dialkylammonium cations, have been prepared. These complexes most probably have a pseudorotaxane geometry.