MilliporeSigma
  • Evidence for the serotonin HTR2A receptor gene as a susceptibility factor in attention deficit hyperactivity disorder (ADHD).
Evidence for the serotonin HTR2A receptor gene as a susceptibility factor in attention deficit hyperactivity disorder (ADHD).
Molecular psychiatry (2000-10-14)
J F Quist, C L Barr, R Schachar, W Roberts, M Malone, R Tannock, V S Basile, J Beitchman, J L Kennedy
ABSTRACT
A recent study demonstrated that treatment of hyperactive mice with psychostimulants and serotonergic agents produced a calming effect that was dependent on serotonergic neurotransmission and was not associated with any changes in extracellular dopamine levels. The complex interaction between the serotonergic and dopaminergic neurotransmitter systems suggests that a balance between the two systems may be necessary for mediating hyperactive behaviour. Defects in serotonin system genes, therefore, may disrupt normal brain serotonin function causing an imbalance between these neurotransmitter systems leading to the development of attention deficit hyperactivity disorder (ADHD). Using the transmission disequilibrium test (TDT), the current study assesses for linkage disequilibrium between polymorphisms in the serotonin HTR2A receptor gene and ADHD. One hundred and fifteen families with a total of 143 children diagnosed with ADHD (DSM-IV) were genotyped for the His452 Tyr and the T102C polymorphisms in the serotonin HTR2A receptor gene. TDT analysis revealed a preferential transmission of the 452Tyr allele to the affected offspring (P = 0.03), suggesting linkage disequilibrium of this polymorphism with ADHD. This may open a new door in ADHD molecular genetics research, expanding the existing view of a catecholaminergic hypothesis to include a serotonergic hypothesis and should help elucidate the complex interplay among the neurotransmitter systems in the etiology of ADHD.