MilliporeSigma
  • Identification and function of adenosine A3 receptor in afferent arterioles.
Identification and function of adenosine A3 receptor in afferent arterioles.
American journal of physiology. Renal physiology (2015-01-23)
Yan Lu, Rui Zhang, Ying Ge, Mattias Carlstrom, Shaohui Wang, Yiling Fu, Liang Cheng, Jin Wei, Richard J Roman, Lei Wang, Xichun Gao, Ruisheng Liu
ABSTRACT
Adenosine plays an important role in regulation of renal microcirculation. All receptors of adenosine, A1, A2A, A2B, and A3, have been found in the kidney. However, little is known about the location and function of the A3 receptor in the kidney. The present study determined the expression and role of A3 receptors in mediating the afferent arteriole (Af-Art) response and studied the interaction of A3 receptors with angiotensin II (ANG II), A1 and A2 receptors on the Af-Art. We found that the A3 receptor expressed in microdissected isolated Af-Art and the mRNA levels of A3 receptor were 59% of A1. In the isolated microperfused Af-Art, A3 receptor agonist IB-MECA did not have a constrictive effect. Activation of A3 receptor dilated the preconstricted Af-Art by norepinephrine and blunted the vasoconstrictive effect of both adenosine A1 receptor activation and ANG II on the Af-Art, respectively. Selective A2 receptor antagonist (both A2A and A2B) had no effect on A3 receptor agonist-induced vasodilation, indicating that the dilatory effect of A3 receptor activation is not mediated by activation of A2 receptor. We conclude that the A3 receptor is expressed in the Af-Art, and activation of the A3 receptor dilates the Af-Art.
Sigma-Aldrich
(E)-4-Amino-2-butenoic acid