MilliporeSigma
  • The familial Parkinsonism gene LRRK2 regulates neurite process morphology.
The familial Parkinsonism gene LRRK2 regulates neurite process morphology.
Neuron (2006-11-23)
David MacLeod, Julia Dowman, Rachel Hammond, Thomas Leete, Keiichi Inoue, Asa Abeliovich
ABSTRACT
Mutations in LRRK2 underlie an autosomal-dominant, inherited form of Parkinson's disease (PD) that mimics the clinical features of the common "sporadic" form of PD. The LRRK2 protein includes putative GTPase, protein kinase, WD40 repeat, and leucine-rich repeat (LRR) domains of unknown function. Here we show that PD-associated LRRK2 mutations display disinhibited kinase activity and induce a progressive reduction in neurite length and branching both in primary neuronal cultures and in the intact rodent CNS. In contrast, LRRK2 deficiency leads to increased neurite length and branching. Neurons that express PD-associated LRRK2 mutations additionally harbor prominent phospho-tau-positive inclusions with lysosomal characteristics and ultimately undergo apoptosis.