MilliporeSigma
  • Synthesis, antitumor activity, and nephrotoxicity of the optical isomers of 2-aminomethylpyrrolidine(1,1-cyclobutane-dicarboxylato)platinum(II).
Synthesis, antitumor activity, and nephrotoxicity of the optical isomers of 2-aminomethylpyrrolidine(1,1-cyclobutane-dicarboxylato)platinum(II).
Journal of pharmaceutical sciences (1991-09-01)
K Morikawa, M Honda, K Endoh, T Matsumoto, K Akamatsu, H Mitsui, M Koizumi
ABSTRACT
The optical isomers of 2-aminomethylpyrrolidine(1,1-cyclobutane- dicarboxylato)platinum(II) (DWA2114, 1), which has potent antitumor activity against various tumors, were synthesized. They were examined for antitumor activity against Colon 26 carcinoma in a sc-iv system, and changes in urinary protein and sugar levels in drug-treated mice were used as an index of nephrotoxicity. In their effect on tumors, (+)-(S)-2-aminomethylpyrrolidine(1,1-cyclobutanedicarboxylato++ +)platinum(II) (6b) was more potent than the enantiomer 6a in that the effective dose of 6b was smaller than that of 6a; but, both drugs exhibited potent antitumor activity. On the other hand, a distinct difference between 6a and 6b was shown in their nephrotoxicity. Isomer 6b induced a great increase in urinary protein and sugar levels in mice, whereas 6a caused no increase in these levels.
Sigma-Aldrich
(S)-(+)-2-(Aminomethyl)pyrrolidine, 97%