MilliporeSigma
  • AURKB-mediated effects on chromatin regulate binding versus release of XIST RNA to the inactive chromosome.
AURKB-mediated effects on chromatin regulate binding versus release of XIST RNA to the inactive chromosome.
The Journal of cell biology (2009-08-26)
Lisa L Hall, Meg Byron, Gayle Pageau, Jeanne B Lawrence
ABSTRACT
How XIST RNA strictly localizes across the inactive X chromosome is unknown; however, prophase release of human XIST RNA provides a clue. Tests of inhibitors that mimic mitotic chromatin modifications implicated an indirect role of PP1 (protein phosphatase 1), potentially via its interphase repression of Aurora B kinase (AURKB), which phosphorylates H3 and chromosomal proteins at prophase. RNA interference to AURKB causes mitotic retention of XIST RNA, unlike other mitotic or broad kinase inhibitors. Thus, AURKB plays an unexpected role in regulating RNA binding to heterochromatin, independent of mechanics of mitosis. H3 phosphorylation (H3ph) was shown to precede XIST RNA release, whereas results exclude H1ph involvement. Of numerous Xi chromatin (chromosomal protein) hallmarks, ubiquitination closely follows XIST RNA retention or release. Surprisingly, H3S10ph staining (but not H3S28ph) is excluded from Xi and is potentially linked to ubiquitination. Results suggest a model of multiple distinct anchor points for XIST RNA. This study advances understanding of RNA chromosome binding and the roles of AURKB and demonstrates a novel approach to manipulate and study XIST RNA.
Sigma-Aldrich
Anti-phospho-Histone H1 Antibody, clone 12D11, culture supernatant, clone 12D11, from mouse