MilliporeSigma
  • Aerosol-OT-gamma-alumina admicelles for the concentration of hydrophobic organic compounds in water.
Aerosol-OT-gamma-alumina admicelles for the concentration of hydrophobic organic compounds in water.
Journal of chromatography. A (2004-07-03)
Tohru Saitoh, Seiichi Matsushima, Masataka Hiraide
ABSTRACT
A novel admicelle composing of a dialkylated anionic surfactant, di-2-ethylhexyl sodium sulfosuccinate (Aerosol-OT, AOT) and gamma-alumina was prepared by mixing them in acidic aqueous solution. The amount of the maximum sorption of AOT on 1 g of alumina at pH 2 was ca. 130 mg. By comparing the fluorescence spectra of N-phenyl-1-naphthylamine in different solvents, the solvent property of AOT-gamma-alumina admicelles was corresponding to that of toluene or diethyl ether. Thus, the AOT-gamma-alumina admicelles had greater hydrophobicity than SDS-gamma-alumina admicelles having similar hydrophobicity to 1-octanol or ethyl acetate. Hydrophobic organic compounds, chlorophenols having more than three chloro substituents, octylphenol, nonylphenol, dibutyl phthalate was almost quantitatively (98% or more) collected onto AOT admicelles composing of 1.5 g gamma-alumina and 150 mg AOT. The greater collection yields rather than those in SDS-admicellar system were ascribable to greater hydrophobicity and stability of AOT admicelles. After the 500-fold concentration, traces (nM) of organic contaminants in water samples were successfully detected with an HPLC having a photometric detector.
Sigma-Aldrich
Dioctyl sulfosuccinate sodium salt, ≥97%