MilliporeSigma
  • Characterization of complexation of poly (N-isopropylacrylamide-co-2-(dimethylamino) ethyl methacrylate) thermoresponsive cationic nanogels with salmon sperm DNA.
Characterization of complexation of poly (N-isopropylacrylamide-co-2-(dimethylamino) ethyl methacrylate) thermoresponsive cationic nanogels with salmon sperm DNA.
International journal of nanomedicine (2009-09-24)
Jim Moselhy, Tasnim Vira, Fei-Fei Liu, Xiao Yu Wu
ABSTRACT
Thermoresponsive cationic nanogel (TCNG) networks based on N-isopropylacrylamide (NIPAM), 2-(dimethylamino)ethyl methacrylate (DMAEMA), and quaternary alkyl ammonium halide salts of DMAEMA (DMAEMAQ) were synthesized by dispersion polymerization technique. The thermoresponsive properties of TCNGs and TCNG-salmon sperm DNA (sasDNA) polyplexes were characterized in aqueous media of various pH and ionic strength. P[NIPAM] and P[NIPAM/DMAEMA] TCNGs exhibited sharp volume phase transition (VPT) in water at critical temperatures (T(c)) of 32 degrees C and 36 degrees C, respectively. Quaternized P[NIPAM/DMAEMAQ] TCNGs did not undergo sharp VPT up to 50 degrees C. The VPT of uncomplexed TCNGs were sensitive to the ionic composition and ionic strength of salts in solution, but were insensitive to pH in the range 5.0 to 7.4. The VPT of P[NIPAM/DMAEMAQ]/sasDNA diminished in magnitude with increasing W(p)/W(d) suggesting greater compaction of the polyplexes. The distinct phase-transition properties of P[NIPAM/DMAEMA]/sasDNA and P[NIPAM/DMAEMAQ]/sasDNA polyplexes were attributed to the condensing capability of polycations and to differences in the spatial distribution of structural charges in quaternized and nonquaternized networks. The findings demonstrate that stable TCNGs can be prepared with controllable responsive properties determined by the nature of the cationic charge incorporated and may have potential as vehicles for DNA delivery.
Sigma-Aldrich
1-Iodopropane, 99%