MilliporeSigma
  • Visualisation of γH2AX foci caused by heavy ion particle traversal; distinction between core track versus non-track damage.
Visualisation of γH2AX foci caused by heavy ion particle traversal; distinction between core track versus non-track damage.
PloS one (2013-08-24)
Nakako Izumi Nakajima, Holly Brunton, Ritsuko Watanabe, Amruta Shrikhande, Ryoichi Hirayama, Naruhiro Matsufuji, Akira Fujimori, Takeshi Murakami, Ryuichi Okayasu, Penny Jeggo, Atsushi Shibata
ABSTRACT
Heavy particle irradiation produces complex DNA double strand breaks (DSBs) which can arise from primary ionisation events within the particle trajectory. Additionally, secondary electrons, termed delta-electrons, which have a range of distributions can create low linear energy transfer (LET) damage within but also distant from the track. DNA damage by delta-electrons distant from the track has not previously been carefully characterised. Using imaging with deconvolution, we show that at 8 hours after exposure to Fe (∼200 keV/µm) ions, γH2AX foci forming at DSBs within the particle track are large and encompass multiple smaller and closely localised foci, which we designate as clustered γH2AX foci. These foci are repaired with slow kinetics by DNA non-homologous end-joining (NHEJ) in G1 phase with the magnitude of complexity diminishing with time. These clustered foci (containing 10 or more individual foci) represent a signature of DSBs caused by high LET heavy particle radiation. We also identified simple γH2AX foci distant from the track, which resemble those arising after X-ray exposure, which we attribute to low LET delta-electron induced DSBs. They are rapidly repaired by NHEJ. Clustered γH2AX foci induced by heavy particle radiation cause prolonged checkpoint arrest compared to simple γH2AX foci following X-irradiation. However, mitotic entry was observed when ∼10 clustered foci remain. Thus, cells can progress into mitosis with multiple clusters of DSBs following the traversal of a heavy particle.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−FITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution