MilliporeSigma
  • Identification of a novel series of tetrahydrodibenzazocines as inhibitors of 17beta-hydroxysteroid dehydrogenase type 3.
Identification of a novel series of tetrahydrodibenzazocines as inhibitors of 17beta-hydroxysteroid dehydrogenase type 3.
Bioorganic & medicinal chemistry letters (2006-01-03)
Brian E Fink, Ashvinikumar V Gavai, John S Tokarski, Bindu Goyal, Raj Misra, Hai-Yun Xiao, S David Kimball, Wen-Ching Han, Derek Norris, Thomas E Spires, Dan You, Marco M Gottardis, Matthew V Lorenzi, Gregory D Vite
ABSTRACT
A novel series of 17beta-hydroxysteroid dehydrogenase type 3 (17beta-HSD3) inhibitors has been identified. These inhibitors, based on a dibenzazocine core, exhibited picomolar to low nanomolar inhibition of 17beta-HSD3 in cell-free enzymatic as well as in cell-based transcriptional reporter assays.
Sigma-Aldrich
5-Bromo-1-indanone, 97%